missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethyl-2-thiourea 97.0+%, TCI America™

Catalog No. D053025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D053025G 25 g
D0530500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D053025G Supplier TCI America Supplier No. D053025G

Chemical Identifiers

CAS 105-55-5
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00004925
InChI Key FLVIGYVXZHLUHP-UHFFFAOYSA-N
Synonym 1,3-diethyl-2-thiourea, n,n'-diethylthiourea, pennzone e, thiourea, n,n'-diethyl, thiate h, n,n'-diethylthiocarbamide, urea, 1,3-diethyl-2-thio, usaf ek-1803, n,n-diethyl-2-thiourea, diethylthiourea
PubChem CID 2735009
ChEBI CHEBI:82448
IUPAC Name 1,3-diethylthiourea
SMILES CCNC(=S)NCC

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 132.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diethyl-2-thiourea
EINECSNumber (2)-1758
RTECSNumber YS9800000
TSCA Yes