missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dicyclohexylimidazolium Tetrafluoroborate 98.0+%, TCI America™
Supplier: TCI America D385025G
Specifications
| 1,3-Dicyclohexylimidazolium Tetrafluoroborate | |
| 174°C | |
| C15H25BF4N2 | |
| 25 g | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 | |
| 320.18 | |
| 320.18 | |
| Crystalline Powder |
| 286014-38-8 | |
| White-Yellow | |
| MFCD04973310 | |
| 1759 | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 1,3-dicyclohexyl-3H-1λâµ-imidazol-1-ylium; tetrafluoroboranuide | |
| 16218169 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 286014-38-8 | |
| 320.18 | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 16218169 | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 |
| C15H25BF4N2 | |
| MFCD04973310 | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| 1,3-dicyclohexyl-3H-1λâµ-imidazol-1-ylium; tetrafluoroboranuide |
Safety and Handling
TSCA : No