missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylimidazolium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7127950
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38501G 1 g
D38505G 5 g
D385025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D38501G Supplier TCI America Supplier No. D38501G
Only null left

Chemical Identifiers

CAS 286014-38-8
Molecular Formula C15H25BF4N2
Molecular Weight (g/mol) 320.18
MDL Number MFCD04973310
InChI Key CQHXJIHJFMBBQA-UHFFFAOYSA-N
Synonym 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt
PubChem CID 16218169
IUPAC Name 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 320.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dicyclohexylimidazolium Tetrafluoroborate