missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dichlorobutane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 134°C |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 127.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,3-Dichlorobutane |
Safety and Handling
| EINECSNumber | (2)-0061 |
| TSCA | Yes |