CAS RN 1190-22-3
IUPAC Name:
1,3-dichlorobutane
Synonyms:
butane, 1,3-dichloro-
Molecular Weight (g/mol):
127.01
Molecular Formula:
C4H8Cl2
InChi Key:
QBGVARBIQGHVKR-UHFFFAOYNA-N
SMILES:
CC(Cl)CCCl
1
–
1
of
1
results
1
1,3-Dichlorobutane 98.0+%, TCI America™
CAS: 1190-22-3 Molecular Formula: C4H8Cl2 Molecular Weight (g/mol): 127.01 MDL Number: MFCD00000870 InChI Key: QBGVARBIQGHVKR-UHFFFAOYNA-N Synonym: butane, 1,3-dichloro,1,3-dichloro-butane,butane,1,3-dichloro,1,3-dichlorobutane PubChem CID: 14480 IUPAC Name: 1,3-dichlorobutane SMILES: CC(Cl)CCCl