missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-iodobenzene 98.0+%, TCI America™

Catalog No. D18665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18665G 5 g
D186625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18665G Supplier TCI America Supplier No. D18665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3032-81-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00001047
InChI Key AATPRMRVLQZEHB-UHFFFAOYSA-N
Synonym 3,5-dichloroiodobenzene, 3,5-dichlorophenyl iodide, benzene, 1,3-dichloro-5-iodo, 1,3-dichloro-5-iodo-benzene, 3,5-dichloro-1-iodobenzene, pubchem3693, acmc-209hev, 3,5-dichloro iodobenzene, 3,5-dichloro-iodobenzene, 3,5-dichloro-iodo-benzene
PubChem CID 76424
IUPAC Name 1,3-dichloro-5-iodobenzene
SMILES ClC1=CC(I)=CC(Cl)=C1

Specifications

Melting Point 58°C
Color Yellow
Boiling Point 127°C
Quantity 5 g
Formula Weight 272.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dichloro-5-iodobenzene