missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-iodobenzene, 99%

Catalog No. p-7021026
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319206 5 g
AAA1319214 25 g
AAA1319222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1319206 Supplier Thermo Scientific Chemicals Supplier No. A1319206

CAS: 3032-81-3 | C6H3Cl2I | 272.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3032-81-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00001047
InChI Key AATPRMRVLQZEHB-UHFFFAOYSA-N
Synonym 3,5-dichloroiodobenzene, 3,5-dichlorophenyl iodide, benzene, 1,3-dichloro-5-iodo, 1,3-dichloro-5-iodo-benzene, 3,5-dichloro-1-iodobenzene, pubchem3693, acmc-209hev, 3,5-dichloro iodobenzene, 3,5-dichloro-iodobenzene, 3,5-dichloro-iodo-benzene
PubChem CID 76424
IUPAC Name 1,3-dichloro-5-iodobenzene
SMILES ClC1=CC(I)=CC(Cl)=C1

Specifications

Melting Point 55°C to 59°C
Boiling Point 127°C (20 mmHg)
Quantity 5 g
Beilstein 2242455
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity 99%
Chemical Name or Material 1,3-Dichloro-5-iodobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-216-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only