missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-fluorobenzene 98.0+%, TCI America™

Catalog No. p-7128072
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40465G Supplier TCI America Supplier No. D40465G
Only null left

Chemical Identifiers

CAS 1435-46-7
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.988
MDL Number MFCD00060652
InChI Key BLWGKIXZAUOECS-UHFFFAOYSA-N
PubChem CID 137002
IUPAC Name 1,3-dichloro-5-fluorobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)F

Specifications

Color Colorless
Boiling Point 162°C
Quantity 5 g
UN Number 1993
Formula Weight 164.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-5-fluorobenzene