missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog No. p-7023924
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1795318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1795318 Supplier Thermo Scientific Chemicals Supplier No. A1795318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1435-48-9 | Molecular Formula: C6H3Cl2F | Molecular Weight: 164.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1435-48-9
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.988
MDL Number MFCD00000580
InChI Key BDJZCCWUSOZUQG-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-fluorobenzene, 2,4-dichlorofluorobenzene, benzene, 2,4-dichloro-1-fluoro, dcfb, benzene,2,4-dichloro-1-fluoro, 2,4-dichloro-1-fluoro-benzene, pubchem2312, acmc-209cqu, 2,4-dichlorofluobenzene, 1,3-dichlorofluorobenzene
PubChem CID 123112
IUPAC Name 2,4-dichloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)F

Specifications

Melting Point -23°C
Density 1.409
Boiling Point 172°C to 174°C
Flash Point 79°C (174°F)
Refractive Index 1.525
Quantity 50 g
UN Number UN2810
Beilstein 2500791
Formula Weight 165
Percent Purity 99%
Chemical Name or Material 1,3-Dichloro-4-fluorobenzene
Show More Show Less
Hazard Category H226-H302-H315-H319-H373
Hazard Statement GHS H Statement
H373-H302-H315-H227
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 406-160-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only