Learn More
1,3-Dichloro-2-propanol, 98+%
CAS: 96-23-1 | C3H6Cl2O | 128.98 g/mol
Supplier: Thermo Scientific Chemicals A160030B
Description
1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.
Solubility
Fully miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 1,3-Dichloro-2-propanol | |
| -4°C | |
| 174°C to 176°C | |
| C3H6Cl2O | |
| ClCH2CH(OH)CH2Cl | |
| 1000 g | |
| 1732063 | |
| 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin | |
| DEWLEGDTCGBNGU-UHFFFAOYSA-N | |
| 1,3-dichloropropan-2-ol | |
| 7289 | |
| 128.99 |
| 96-23-1 | |
| 1.366 | |
| 85°C (185°F) | |
| 1.483 | |
| MFCD00000951 | |
| UN2750 | |
| 14,3074 | |
| Fully miscible in water. | |
| OC(CCl)CCl | |
| 128.98 | |
| CHEBI:18917 | |
| ≥98% |
Chemical Identifiers
| 96-23-1 | |
| 128.98 | |
| DEWLEGDTCGBNGU-UHFFFAOYSA-N | |
| 7289 | |
| 1,3-dichloropropan-2-ol |
| C3H6Cl2O | |
| MFCD00000951 | |
| 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin | |
| CHEBI:18917 | |
| OC(CCl)CCl |
Safety and Handling
GHS H Statement
H301-H311-H350-H227
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
Combustible liquid.
P201-P202-P210-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
H227-H301-H312-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROPROPANOL-2
EINECSNumber : 202-491-9
RTECSNumber : UB1400000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only