missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodobenzene 98.0+%, TCI America™

Catalog No. p-7127812
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35935G 5 g
D359325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35935G Supplier TCI America Supplier No. D35935G

Chemical Identifiers

CAS 19230-28-5
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00060658
InChI Key ZMPGXSFTXBOKFM-UHFFFAOYSA-N
Synonym 2,6-dichloroiodobenzene, benzene, 1,3-dichloro-2-iodo, pubchem9581, acmc-1c3f6, ksc494i7b, 1,3-dichloro-2-iodo-benzene, #, benzene,1,3-dichloro-2-iodo, attercop-chm at115975
PubChem CID 140498
IUPAC Name 1,3-dichloro-2-iodobenzene
SMILES ClC1=CC=CC(Cl)=C1I

Specifications

Melting Point 64°C
Quantity 5 g
Formula Weight 272.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dichloro-2-iodobenzene