missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodobenzene, 98%

Catalog No. p-7036904
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5536503 1 g
AAH5536506 5 g
AAH5536514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5536503 Supplier Thermo Scientific Chemicals Supplier No. H5536503
Only null left

CAS: 19230-28-5 | C6H3Cl2I | 272.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19230-28-5
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00060658
InChI Key ZMPGXSFTXBOKFM-UHFFFAOYSA-N
PubChem CID 140498
IUPAC Name 1,3-dichloro-2-iodobenzene
SMILES ClC1=CC=CC(Cl)=C1I

Specifications

Melting Point 64°C to 67°C
Quantity 1 g
Sensitivity Light sensitive
Synonym 2,6-dichloroiodobenzene, benzene, 1,3-dichloro-2-iodo, pubchem9581, acmc-1c3f6, ksc494i7b, 1,3-dichloro-2-iodo-benzene, #, benzene,1,3-dichloro-2-iodo, attercop-chm at115975
Formula Weight 272.9
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only