missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetramethyldisiloxane 97.0+%, TCI America™

Catalog No. p-7127239
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
D23345G 5 g
D233425G 25 g
2 options

Catalog No. D23345G

Supplier: TCI America D23345G

Only null left

Chemical Identifiers

CAS 2401-73-2
Molecular Formula C4H12Cl2OSi2
Molecular Weight (g/mol) 203.209
MDL Number MFCD00018088
InChI Key DMEXFOUCEOWRGD-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetramethyldisiloxane, 1,3-dichlorotetramethyldisiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetramethyl, chloro chlorodimethylsilyl oxy dimethylsilane, 1,1,3,3-tetramethyl-1,3-dichlorodisiloxane, acmc-209g7w, oxybis chlorodimethylsilane, sym-dichlorotetramethyldisiloxane, dmexfouceowrgd-uhfffaoysa, 1,3-dichlorotetramethyl disiloxane
PubChem CID 75461
IUPAC Name chloro-[chloro(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)Cl)Cl

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 2986
Formula Weight 203.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetramethyldisiloxane
EINECSNumber (7)-0472
TSCA Yes