accessibility menu, dialog, popup

UserName

H3CSiCH3ClOSiCH3CH3Cl

CAS RN 2401-73-2

IUPAC Name: chloro[(chlorodimethylsilyl)oxy]dimethylsilane
Synonyms: chloro[(chlorodimethylsilyl)oxy]dimethylsilane
Molecular Weight (g/mol): 203.21
Molecular Formula: C4H12Cl2OSi2
InChi Key: DMEXFOUCEOWRGD-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)O[Si](C)(C)Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results