missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibutylthiourea 97.0+%, TCI America™

Catalog No. D030125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030125G 25 g
D0301500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D030125G Supplier TCI America Supplier No. D030125G

Chemical Identifiers

CAS 109-46-6
Molecular Formula C9H20N2S
Molecular Weight (g/mol) 188.333
MDL Number MFCD00004926
InChI Key KFFQABQEJATQAT-UHFFFAOYSA-N
Synonym 1,3-dibutyl-2-thiourea, n,n'-dibutylthiourea, thiourea, n,n'-dibutyl, pennzone b, n,n'-di-n-butylthiourea, thiate u, 1,3-di-n-butyl-2-thiourea, urea, 1,3-dibutyl-2-thio, usaf ek-2138, urea, 1,3-di-n-butyl-2-thio
PubChem CID 2723622
IUPAC Name 1,3-dibutylthiourea
SMILES CCCCNC(=S)NCCCC

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 128°C
Quantity 25 g
Formula Weight 188.33
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibutylthiourea
EINECSNumber (2)-1760
RTECSNumber YS8400000
TSCA Yes