missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromotetrafluorobenzene 98.0+%, TCI America™

Catalog No. p-7126748
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14921G 1 g
D14925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14921G Supplier TCI America Supplier No. D14921G
Only null left

Chemical Identifiers

CAS 1559-87-1
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.87
MDL Number MFCD00031533
InChI Key UCWKDDQEZQRGDR-UHFFFAOYSA-N
PubChem CID 73793
IUPAC Name 1,3-dibromo-2,4,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(Br)=C(F)C(Br)=C1F

Specifications

Color Yellow
Quantity 1 g
Synonym 1,3-dibromotetrafluorobenzene, benzene, 1,3-dibromotetrafluoro, benzene, 1,3-dibromo-2,4,5,6-tetrafluoro, 1,3-dibromo-2,4,5,6-tetrafluoro-benzene, benzene,1,3-dibromo-2,4,5,6-tetrafluoro, acmc-1byn7, benzene,3-dibromotetrafluoro, 1,3-dibromotetrafluorobenzene, 98+%
Formula Weight 307.87
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromotetrafluorobenzene