missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromotetrafluorobenzene, 98+%

Catalog No. p-7023635
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1739206 5 g
AAA1739214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1739206 Supplier Thermo Scientific Chemicals Supplier No. A1739206

CAS: 1559-87-1 | C6Br2F4 | 307.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1559-87-1
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.87
MDL Number MFCD00031533
InChI Key UCWKDDQEZQRGDR-UHFFFAOYSA-N
PubChem CID 73793
IUPAC Name 1,3-dibromo-2,4,5,6-tetrafluorobenzene
SMILES FC1=C(F)C(Br)=C(F)C(Br)=C1F

Specifications

Density 2.18
Boiling Point 85°C to 88°C (18 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.516
Quantity 5 g
Synonym 1,3-dibromotetrafluorobenzene, benzene, 1,3-dibromotetrafluoro, benzene, 1,3-dibromo-2,4,5,6-tetrafluoro, 1,3-dibromo-2,4,5,6-tetrafluoro-benzene, benzene,1,3-dibromo-2,4,5,6-tetrafluoro, acmc-1byn7, benzene,3-dibromotetrafluoro, 1,3-dibromotetrafluorobenzene, 98+%
Formula Weight 307.87
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibromotetrafluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-326-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only