missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane, 98%

Catalog No. p-7022080
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA146870B 1000 g
AAA146870I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA146870B Supplier Thermo Scientific Chemicals Supplier No. A146870B
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

1,3-Dibromopropane, 98%, C3H6Br2, CAS Number-109-64-8, 1,3-dibromo propane, 1,3-dibromo-propane, .alpha.,.gamma.-dibromopropane, alpha,gamma-dibromopropane, ccris 6711, .omega.,.omega.'-dibromopropane, propane, 1,3-dibromo, trimethylene bromide, trimethylene dibromide, unii-yqr3048ix9, 5000g, 635662 | CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol

1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 109-64-8
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000255
InChI Key VEFLKXRACNJHOV-UHFFFAOYSA-N
Synonym trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
PubChem CID 8001
IUPAC Name 1,3-dibromopropane
SMILES BrCCCBr

Specifications

Melting Point -34°C
Density 1.99
Boiling Point 167°C
Flash Point 54°C (130°F)
Assay Percent Range 98%
Refractive Index 1.523
Linear Formula Br(CH2)3Br
Quantity 1000 g
UN Number UN1993
Beilstein 635662
Merck Index 14,9713
Solubility Information Miscible with water.
Formula Weight 201.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dibromopropane
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-690-3
RTECSNumber TX8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only