missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane, 98%, stabilized

Catalog No. p-102391
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155041000 100 mL Glass bottle
AC155045000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155041000 Supplier Thermo Scientific Chemicals Supplier No. 155041000
Only null left

CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-64-8
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000255
InChI Key VEFLKXRACNJHOV-UHFFFAOYSA-N
Synonym trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
PubChem CID 8001
IUPAC Name 1,3-dibromopropane
SMILES BrCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -34.0°C
Color Colorless to Yellow
Density 1.9890g/mL
Boiling Point 167.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5215 to 1.5235
Linear Formula Br(CH2)3Br
Quantity 100 mL
Beilstein 01, 110
Fieser 13,313
Specific Gravity 1.989
Merck Index 15, 9884
Solubility Information Solubility in water: 1.7g/l (20°C). Other solubilities: soluble in organic solvents
Stabilizer 400ppm butylene oxide
Formula Weight 201.89
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dibromopropane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 203-690-3
RTECSNumber TX8575000
TSCA TSCA

RUO – Research Use Only