missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromoazulene 98.0+%, TCI America™

Catalog No. D4662200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4662200MG 200 mg
D46621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4662200MG Supplier TCI America Supplier No. D4662200MG

Chemical Identifiers

CAS 14658-95-8
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.966
InChI Key OEXLRDNVSRMWHO-UHFFFAOYSA-N
PubChem CID 139802
IUPAC Name 1,3-dibromoazulene
SMILES C1=CC=C2C(=C(C=C2Br)Br)C=C1

Specifications

Melting Point 90°C
Color Black-Blue
Quantity 200 mg
Formula Weight 285.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromoazulene