accessibility menu, dialog, popup

UserName

BrBr

CAS RN 14658-95-8

IUPAC Name: 1,3-dibromoazulene
Synonyms: 1,3-dibromoazulene
Molecular Weight (g/mol): 285.97
Molecular Formula: C10H6Br2
InChi Key: OEXLRDNVSRMWHO-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C2C=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results