missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-n-octylbenzene 98.0+%, TCI America™

$130.92 - $438.93

Chemical Identifiers

CAS 75894-99-4
Molecular Formula C14H20Br2
Molecular Weight (g/mol) 348.122
InChI Key OHOXSSBAVJERNK-UHFFFAOYSA-N
PubChem CID 101748485
IUPAC Name 1,3-dibromo-5-octylbenzene
SMILES CCCCCCCCC1=CC(=CC(=C1)Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D52571G
View Documents
TCI America
D52571G
1 g N/A
Only null left
 
D52575G
View Documents
TCI America
D52575G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

75894-99-4
348.122
101748485
CCCCCCCCC1=CC(=CC(=C1)Br)Br
C14H20Br2
OHOXSSBAVJERNK-UHFFFAOYSA-N
1,3-dibromo-5-octylbenzene

Specifications

75894-99-4
1 g
CCCCCCCCC1=CC(=CC(=C1)Br)Br
348.122
348.12
Liquid
C14H20Br2
OHOXSSBAVJERNK-UHFFFAOYSA-N
1,3-dibromo-5-octylbenzene
101748485
≥98.0% (GC)
1,3-Dibromo-5-n-octylbenzene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents