missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-5-n-octylbenzene 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 348.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,3-Dibromo-5-n-octylbenzene |
Safety and Handling
| TSCA | No |