missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-iodobenzene 98.0+%, TCI America™

Catalog No. D46911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46911G 1 g
D46915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46911G Supplier TCI America Supplier No. D46911G
Only null left

Chemical Identifiers

CAS 19752-57-9
Molecular Formula C6H3Br2I
Molecular Weight (g/mol) 361.80
MDL Number MFCD07778996
InChI Key KARUMYWDGQTPFL-UHFFFAOYSA-N
PubChem CID 11068437
IUPAC Name 1,3-dibromo-5-iodobenzene
SMILES BrC1=CC(I)=CC(Br)=C1

Specifications

Melting Point 123°C
Quantity 1 g
Formula Weight 361.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-5-iodobenzene