missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-hexylbenzene 95.0+%, TCI America™

Catalog No. p-7128464
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47891G 1 g
D47895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47891G Supplier TCI America Supplier No. D47891G

Chemical Identifiers

CAS 75894-97-2
Molecular Formula C12H16Br2
Molecular Weight (g/mol) 320.068
InChI Key LAKHKLCGQAELGL-UHFFFAOYSA-N
PubChem CID 85996911
IUPAC Name 1,3-dibromo-5-hexylbenzene
SMILES CCCCCCC1=CC(=CC(=C1)Br)Br

Specifications

Color Yellow
Boiling Point 142°C
Quantity 1 g
Formula Weight 320.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromo-5-hexylbenzene