accessibility menu, dialog, popup

UserName

H3CBrBr

CAS RN 75894-97-2

IUPAC Name: 1,3-dibromo-5-hexylbenzene
Synonyms: 1,3-dibromo-5-hexylbenzene
Molecular Weight (g/mol): 320.07
Molecular Formula: C12H16Br2
InChi Key: LAKHKLCGQAELGL-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results