missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-dodecylbenzene 97.0+%, TCI America™

Catalog No. p-7128721
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52741G 1 g
D52745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52741G Supplier TCI America Supplier No. D52741G
Only null left

Chemical Identifiers

CAS 231606-27-2
Molecular Formula C18H28Br2
Molecular Weight (g/mol) 404.23
InChI Key LBSQLYHNNHYVBN-UHFFFAOYSA-N
PubChem CID 91153088
IUPAC Name 1,3-dibromo-5-dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC(=CC(=C1)Br)Br

Specifications

Melting Point 53°C
Quantity 1 g
Formula Weight 404.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Dibromo-5-dodecylbenzene
TSCA No
Recommended Storage Refrigerator