missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-5-dodecylbenzene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 53°C |
| Quantity | 5 g |
| Formula Weight | 404.23 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 1,3-Dibromo-5-dodecylbenzene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |