missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-chlorobenzene 96.0+%, TCI America™

Catalog No. p-7127743
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34655G 5 g
D346525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34655G Supplier TCI America Supplier No. D34655G

Chemical Identifiers

CAS 14862-52-3
Molecular Formula C6H3Br2Cl
Molecular Weight (g/mol) 270.348
MDL Number MFCD00070765
InChI Key FNKCOUREFBNNHG-UHFFFAOYSA-N
Synonym 3,5-dibromochlorobenzene, 1-chloro-3,5-dibromobenzene, benzene, 1,3-dibromo-5-chloro, pubchem3634, ambsclk-224, intermediates-zcf02139, ksc181m3d, 1,3-dibromo-5-chloro-benzene, tpc-i027, fnkcourefbnnhg-uhfffaoysa
PubChem CID 84676
IUPAC Name 1,3-dibromo-5-chlorobenzene
SMILES C1=C(C=C(C=C1Br)Br)Cl

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 256°C
Formula Weight 270.35
Percent Purity ≥96.0% (GC)
Chemical Name or Material 1,3-Dibromo-5-chlorobenzene