accessibility menu, dialog, popup

UserName

CAS RN 14862-52-3

IUPAC Name: 1,3-dibromo-5-chlorobenzene
Synonyms: 1,3-dibromo-5-chlorobenzene
Molecular Weight (g/mol): 270.35
Molecular Formula: C6H3Br2Cl
InChi Key: FNKCOUREFBNNHG-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)