missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-chlorobenzene, 98%, Thermo Scientific Chemicals

Catalog No. p-7028221
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2541114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2541114 Supplier Thermo Scientific Chemicals Supplier No. B2541114

CAS: 14862-52-3 | Molecular Formula: C6H3Br2Cl | Molecular Weight: 270.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14862-52-3
Molecular Formula C6H3Br2Cl
Molecular Weight (g/mol) 270.348
MDL Number MFCD00070765
InChI Key FNKCOUREFBNNHG-UHFFFAOYSA-N
Synonym 3,5-dibromochlorobenzene, 1-chloro-3,5-dibromobenzene, benzene, 1,3-dibromo-5-chloro, pubchem3634, ambsclk-224, intermediates-zcf02139, ksc181m3d, 1,3-dibromo-5-chloro-benzene, tpc-i027, fnkcourefbnnhg-uhfffaoysa
PubChem CID 84676
IUPAC Name 1,3-dibromo-5-chlorobenzene
SMILES C1=C(C=C(C=C1Br)Br)Cl

Specifications

Melting Point 95°C to 98°C
Quantity 25 g
Formula Weight 270.35
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5-chlorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only