Learn More
1,3-Dibromo-5,5-dimethylhydantoin, 98%
CAS: 77-48-5 | C5H6Br2N2O2 | 285.923 g/mol
Supplier: Thermo Scientific Chemicals A155100E
Description
It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.
Solubility
Soluble in Alcohol, Ether, Benzene. Insoluble in Water.
Notes
Moisture sensitive. Store away from oxidizing agents, reducing agents, flammable substances, organic materials, metal powders & moisture.
Specifications
| 1,3-Dibromo-5,5-dimethylhydantoin | |
| ∼198°C (decomposition) | |
| C5H6Br2N2O2 | |
| 2500 g | |
| 146024 | |
| 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz | |
| VRLDVERQJMEPIF-UHFFFAOYSA-N | |
| 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione | |
| 6479 | |
| 98% |
| 77-48-5 | |
| 155°C (311°F) | |
| MFCD00003189 | |
| UN3085 | |
| Moisture sensitive | |
| Soluble in Alcohol,Ether,Benzene. Insoluble in Water. | |
| CC1(C(=O)N(C(=O)N1Br)Br)C | |
| 285.923 | |
| 285.93 |
Chemical Identifiers
| 77-48-5 | |
| 285.923 | |
| VRLDVERQJMEPIF-UHFFFAOYSA-N | |
| 6479 | |
| CC1(C(=O)N(C(=O)N1Br)Br)C |
| C5H6Br2N2O2 | |
| MFCD00003189 | |
| 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz | |
| 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione |
Safety and Handling
GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P220-P221-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H272-H302-H314
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber : 201-030-9
RTECSNumber : MU0686000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only