missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5,5-dimethylhydantoin, 98%

Catalog No. p-7022639
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1551022 100 g
AAA1551036 500 g
AAA155100E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1551022 Supplier Thermo Scientific Chemicals Supplier No. A1551022

CAS: 77-48-5 | C5H6Br2N2O2 | 285.923 g/mol

It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for drinking water purification, recreational water treatment, as a bleaching agent in pulp and paper mills, and for treating industrial/commercial water cooling systems. It is an efficient and selective agent for the oxidation of thiols to disulfides in solution or under solvent-free conditions.

Solubility
Soluble in Alcohol, Ether, Benzene. Insoluble in Water.

Notes
Moisture sensitive. Store away from oxidizing agents, reducing agents, flammable substances, organic materials, metal powders & moisture.

Chemical Identifiers

CAS 77-48-5
Molecular Formula C5H6Br2N2O2
Molecular Weight (g/mol) 285.923
MDL Number MFCD00003189
InChI Key VRLDVERQJMEPIF-UHFFFAOYSA-N
Synonym 1,3-dibromo-5,5-dimethylhydantoin, dibromantin, dibromantine, take charge orange, dbdmh, n,n'-dibromodimethylhydantoin, 5,5-dimethyl-1,3-dibromohydantoin, 2,4-imidazolidinedione, 1,3-dibromo-5,5-dimethyl, 1,3-dibromo-5,5-dimethyl-2,4-imidazolidinedione, unii-v9r5f9i7mz
PubChem CID 6479
IUPAC Name 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)N(C(=O)N1Br)Br)C

Specifications

Melting Point ∼198°C (decomposition)
Flash Point 155°C (311°F)
Quantity 100 g
UN Number UN3085
Beilstein 146024
Sensitivity Moisture sensitive
Solubility Information Soluble in Alcohol,Ether,Benzene. Insoluble in Water.
Formula Weight 285.93
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-5,5-dimethylhydantoin
Hazard Category H272-H302-H314
Hazard Statement GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber 201-030-9
RTECSNumber MU0686000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only