missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-3-methylbutane, 98%

Catalog No. AAB2046806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2046806 5 g
AAB2046814 25 g
AAB2046822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2046806 Supplier Thermo Scientific Chemicals Supplier No. B2046806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24443-15-0 | C5H10Br2 | 229.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24443-15-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.94
MDL Number MFCD00060773
InChI Key XMJFMQSXQQUJTQ-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo-3-methyl, acmc-20ap4g, 2,4-dibromo-2-methylbutane
PubChem CID 141113
IUPAC Name 1,3-dibromo-3-methylbutane
SMILES CC(C)(Br)CCBr

Specifications

Density 1.69
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 229.95
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-3-methylbutane
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only