missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-2-propanol (stabilized with Copper chip) 95.0+%, TCI America™

Catalog No. p-7126076
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D018725ML 25 mL
D0187100ML 100 mL
D0187500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D018725ML Supplier TCI America Supplier No. D018725ML
Only null left

Chemical Identifiers

CAS 96-21-9
Molecular Formula C3H6Br2O
Molecular Weight (g/mol) 217.89
MDL Number MFCD00000216
InChI Key KIHQZLPHVZKELA-UHFFFAOYSA-N
Synonym 1,3-dibromo-2-propanol, 2-propanol, 1,3-dibromo, 1,3-dibromohydrin, glycerol 1,3-dibromohydrin, 2-hydroxy-1,3-dibromopropane, 1,3-dibromo-2-hydroxypropane, alpha-dibromohydrin, .alpha.-dibromohydrin, glycerol alpha,gamma-dibromohydrin, glycerol alpha,gamma-dibromohydrine
PubChem CID 7287
IUPAC Name 1,3-dibromopropan-2-ol
SMILES OC(CBr)CBr

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 mL
UN Number 1987
Formula Weight 217.89
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromo-2-propanol (stabilized with Copper chip)
EINECSNumber (2)-2002
RTECSNumber UB0200000
TSCA Yes
Recommended Storage Refrigerator