missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-2-propanol, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Color | Brown or Colorless to Yellow |
| Density | 2.1300g/mL |
| Boiling Point | 219°C |
| Flash Point | 46°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5500 to 1.554 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Suspected of causing genetic defects. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN |
| EINECSNumber | 202-489-8 |
| RTECSNumber | UB0200000 |
| TSCA | TSCA |
RUO – Research Use Only