missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-2-iodobenzene 98.0+%, TCI America™
$113.59 - $333.05
Chemical Identifiers
| CAS | 19821-80-8 |
|---|---|
| Molecular Formula | C6H3Br2I |
| Molecular Weight (g/mol) | 361.80 |
| MDL Number | MFCD00192666 |
| InChI Key | OIFRMZVTJQAPIF-UHFFFAOYSA-N |
| PubChem CID | 4403794 |
| IUPAC Name | 1,3-dibromo-2-iodobenzene |
| SMILES | BrC1=CC=CC(Br)=C1I |
Chemical Identifiers
| 19821-80-8 | |
| 361.80 | |
| OIFRMZVTJQAPIF-UHFFFAOYSA-N | |
| 1,3-dibromo-2-iodobenzene |
| C6H3Br2I | |
| MFCD00192666 | |
| 4403794 | |
| BrC1=CC=CC(Br)=C1I |
Specifications
| 19821-80-8 | |
| White-Yellow | |
| MFCD00192666 | |
| OIFRMZVTJQAPIF-UHFFFAOYSA-N | |
| 1,3-dibromo-2-iodobenzene | |
| 4403794 | |
| ≥98.0% (GC) | |
| 1,3-Dibromo-2-iodobenzene |
| 100°C | |
| C6H3Br2I | |
| 1 g | |
| BrC1=CC=CC(Br)=C1I | |
| 361.80 | |
| 361.80 | |
| Crystalline Powder |
Safety and Handling
TSCA : No