missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibenzoyloxybenzene 98.0+%, TCI America™

Catalog No. p-7128694
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52075G 5 g
D520725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52075G Supplier TCI America Supplier No. D52075G
Only null left

Chemical Identifiers

CAS 94-01-9
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00016576
InChI Key SUQGLJRNDJRARS-UHFFFAOYSA-N
Synonym 1,3-dibenzoyloxybenzene, resorcinol dibenzoate, 1,3-benzenediol, dibenzoate, 1,3-phenylene dibenzoate, m-phenylene dibenzoate, resorcinol, dibenzoate, 1,3-bis benzoyloxy benzene, 3-benzoyloxy phenyl benzoate, 1,3-benzenediol, 1,3-dibenzoate, 1,3-phenylenedibenzoate
PubChem CID 66742
IUPAC Name 3-(benzoyloxy)phenyl benzoate
SMILES O=C(OC1=CC(OC(=O)C2=CC=CC=C2)=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 318.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibenzoyloxybenzene
RTECSNumber VH0590000
TSCA Yes