accessibility menu, dialog, popup

UserName

CAS RN 94-01-9

IUPAC Name: 3-(benzoyloxy)phenyl benzoate
Synonyms: 1,3-benzenediol, dibenzoate
Molecular Weight (g/mol): 318.33
Molecular Formula: C20H14O4
InChi Key: SUQGLJRNDJRARS-UHFFFAOYSA-N
SMILES: O=C(OC1=CC(OC(=O)C2=CC=CC=C2)=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)