Learn More
1,3-Dibenzoylbenzene, 98+%
CAS: 3770-82-9 | C20H14O2 | 286.33 g/mol
Supplier: Thermo Scientific Chemicals A1276814
Description
1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.
Solubility
Sparingly Soluble (4.0E-3 g/L) (25°C).
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
| 1,3-Dibenzoylbenzene | |
| 98°C to 99°C | |
| MFCD00014086 | |
| 1973337 | |
| Sparingly Soluble (4.0E-3g/L) (25°C). | |
| C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)C3=CC=CC=C3 | |
| 286.33 | |
| 286.33 |
| 3770-82-9 | |
| C20H14O2 | |
| 25 g | |
| 1,3-dibenzoylbenzene, 1,3-phenylenebis phenylmethanone, m-dibenzoylbenzene, methanone, 1,3-phenylenebis phenyl, 3-benzoylphenyl phenyl methanone, m-dibenzoyl benzene, methanone, 1,1'-1,3-phenylene bis 1-phenyl, phenyl 3-phenylcarbonyl phenyl ketone | |
| MJQHDSIEDGPFAM-UHFFFAOYSA-N | |
| (3-benzoylphenyl)-phenylmethanone | |
| 77388 | |
| ≥98% |
Chemical Identifiers
| 3770-82-9 | |
| 286.33 | |
| MJQHDSIEDGPFAM-UHFFFAOYSA-N | |
| 77388 | |
| C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)C3=CC=CC=C3 |
| C20H14O2 | |
| MFCD00014086 | |
| 1,3-dibenzoylbenzene, 1,3-phenylenebis phenylmethanone, m-dibenzoylbenzene, methanone, 1,3-phenylenebis phenyl, 3-benzoylphenyl phenyl methanone, m-dibenzoyl benzene, methanone, 1,1'-1,3-phenylene bis 1-phenyl, phenyl 3-phenylcarbonyl phenyl ketone | |
| (3-benzoylphenyl)-phenylmethanone |
Safety and Handling
EINECSNumber : 223-210-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only