missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibenzoylbenzene, 98+%

Catalog No. AAA1276806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276806 5 g
AAA1276814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1276806 Supplier Thermo Scientific Chemicals Supplier No. A1276806

CAS: 3770-82-9 | C20H14O2 | 286.33 g/mol

1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibenzoylbenzene is suitable reagent used in the synthesis of 1,3-bis(1-phenylvinyl)benzene (MDDPE). It may be used in the synthesis of polyanionic spin-quintet molecular clusters and ground state undecet hydrocarbon with 10 parallel spins.

Solubility
Sparingly Soluble (4.0E-3 g/L) (25°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 3770-82-9
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00014086
InChI Key MJQHDSIEDGPFAM-UHFFFAOYSA-N
Synonym 1,3-dibenzoylbenzene, 1,3-phenylenebis phenylmethanone, m-dibenzoylbenzene, methanone, 1,3-phenylenebis phenyl, 3-benzoylphenyl phenyl methanone, m-dibenzoyl benzene, methanone, 1,1'-1,3-phenylene bis 1-phenyl, phenyl 3-phenylcarbonyl phenyl ketone
PubChem CID 77388
IUPAC Name (3-benzoylphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 98°C to 99°C
Quantity 5 g
Beilstein 1973337
Solubility Information Sparingly Soluble (4.0E-3g/L) (25°C).
Formula Weight 286.33
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibenzoylbenzene
EINECSNumber 223-210-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only