missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diaminopropane 98.0+%, TCI America™
Supplier: TCI America D0114500ML
Specifications
| 1,3-Diaminopropane | |
| Yellow | |
| C3H10N2 | |
| 500 mL | |
| 1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg | |
| C(CN)CN | |
| 74.127 | |
| CHEBI:15725 | |
| ≥98.0% (GC) |
| 109-76-2 | |
| 133°C | |
| MFCD00008228 | |
| 2734 | |
| XFNJVJPLKCPIBV-UHFFFAOYSA-N | |
| propane-1,3-diamine | |
| 428 | |
| 74.13 | |
| Liquid |
Chemical Identifiers
| 109-76-2 | |
| 74.127 | |
| XFNJVJPLKCPIBV-UHFFFAOYSA-N | |
| 428 | |
| propane-1,3-diamine |
| C3H10N2 | |
| MFCD00008228 | |
| 1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg | |
| CHEBI:15725 | |
| C(CN)CN |
Safety and Handling
EINECSNumber : (2)-0149
RTECSNumber : TX6825000
TSCA : Yes