missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diaminopropane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 133°C |
| Quantity | 25 mL |
| UN Number | 2734 |
| Formula Weight | 74.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,3-Diaminopropane |
Safety and Handling
| EINECSNumber | (2)-0149 |
| RTECSNumber | TX6825000 |
| TSCA | Yes |