missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane-N,N'-diacetic Acid 98.0+%, TCI America™

Catalog No. P11155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11155G 5 g
P111525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11155G Supplier TCI America Supplier No. P11155G

Chemical Identifiers

CAS 112041-05-1
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.199
MDL Number MFCD00142534
InChI Key RKPHYUBLENEQNT-UHFFFAOYSA-N
PubChem CID 414255
IUPAC Name 2-[3-(carboxymethylamino)propylamino]acetic acid
SMILES C(CNCC(=O)O)CNCC(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Synonym 1,3-Propylenediamine-N,N′C-diacetic Acid
Formula Weight 190.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane-N,N′-diacetic Acid