accessibility menu, dialog, popup

UserName

NHOOHNHOOH

CAS RN 112041-05-1

IUPAC Name: 2-({3-[(carboxymethyl)amino]propyl}amino)acetic acid
Synonyms: ({3-[(carboxymethyl)amino]propyl}amino)acetic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C7H14N2O4
InChi Key: RKPHYUBLENEQNT-UHFFFAOYSA-N
SMILES: OC(=O)CNCCCNCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results