missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane Dihydroiodide 98.0+%, TCI America™

Catalog No. D50915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50915G 5 g
D50911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50915G Supplier TCI America Supplier No. D50915G

Chemical Identifiers

CAS 120675-53-8
Molecular Formula C3H12I2N2
Molecular Weight (g/mol) 329.952
InChI Key KFXBDBPOGBBVMC-UHFFFAOYSA-N
Synonym 1,3-Propanediamine Dihydroiodide, Trimethylenediamine Dihydroiodide
PubChem CID 129655464
IUPAC Name propane-1,3-diamine;dihydroiodide
SMILES C(CN)CN.I.I

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 329.95
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane Dihydroiodide