accessibility menu, dialog, popup

UserName

CAS RN 120675-53-8

NH2H2NHIHI

CAS RN 120675-53-8

IUPAC Name: propane-1,3-diamine dihydroiodide
Synonyms: α,ω-propanediamine dihydroiodide
Molecular Weight (g/mol): 329.95
Molecular Formula: C3H12I2N2
InChi Key: KFXBDBPOGBBVMC-UHFFFAOYSA-N
SMILES: I.I.NCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results