missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminopropane Dihydrobromide 98.0+%, TCI America™

Catalog No. p-7128650
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50901G 1 g
D50905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50901G Supplier TCI America Supplier No. D50901G
Only null left

Chemical Identifiers

CAS 18773-03-0
Molecular Formula C3H12Br2N2
Molecular Weight (g/mol) 235.951
InChI Key FSAFLEKOBQFCHY-UHFFFAOYSA-N
Synonym 1,3-Propanediamine Dihydrobromide, Trimethylenediamine Dihydrobromide
PubChem CID 88779942
IUPAC Name propane-1,3-diamine;dihydrobromide
SMILES C(CN)CN.Br.Br

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 235.95
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane Dihydrobromide