accessibility menu, dialog, popup

UserName

CAS RN 18773-03-0

IUPAC Name: propane-1,3-diamine dihydrobromide
Synonyms: α,ω-propanediamine dihydrobromide
Molecular Weight (g/mol): 235.95
Molecular Formula: C3H12Br2N2
InChi Key: FSAFLEKOBQFCHY-UHFFFAOYSA-N
SMILES: Br.Br.NCCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results