missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diallylurea 98.0+%, TCI America™
$300.11 - $887.29
Chemical Identifiers
| CAS | 1801-72-5 |
|---|---|
| Molecular Formula | C7H12N2O |
| Molecular Weight (g/mol) | 140.186 |
| MDL Number | MFCD00008639 |
| InChI Key | QRWVOJLTHSRPOA-UHFFFAOYSA-N |
| Synonym | 1,3-diallylurea, n,n'-diallylurea, urea, n,n'-di-2-propenyl, urea, 1,3-diallyl, unii-lxm0xy469i, diallyl urea, lxm0xy469i, urea,n,n'-di-2-propen-1-yl, diallylurea, sinapoline |
| PubChem CID | 74537 |
| IUPAC Name | 1,3-bis(prop-2-enyl)urea |
| SMILES | C=CCNC(=O)NCC=C |
Chemical Identifiers
| 1801-72-5 | |
| 140.186 | |
| QRWVOJLTHSRPOA-UHFFFAOYSA-N | |
| 74537 | |
| C=CCNC(=O)NCC=C |
| C7H12N2O | |
| MFCD00008639 | |
| 1,3-diallylurea, n,n'-diallylurea, urea, n,n'-di-2-propenyl, urea, 1,3-diallyl, unii-lxm0xy469i, diallyl urea, lxm0xy469i, urea,n,n'-di-2-propen-1-yl, diallylurea, sinapoline | |
| 1,3-bis(prop-2-enyl)urea |
Specifications
| 1801-72-5 | |
| White | |
| C7H12N2O | |
| 1 g | |
| QRWVOJLTHSRPOA-UHFFFAOYSA-N | |
| 1,3-bis(prop-2-enyl)urea | |
| 74537 | |
| ≥98.0% (GC) | |
| 1,3-Diallylurea |
| 98°C | |
| 56°C | |
| MFCD00008639 | |
| 1,3-diallylurea, n,n'-diallylurea, urea, n,n'-di-2-propenyl, urea, 1,3-diallyl, unii-lxm0xy469i, diallyl urea, lxm0xy469i, urea,n,n'-di-2-propen-1-yl, diallylurea, sinapoline | |
| C=CCNC(=O)NCC=C | |
| 140.186 | |
| 140.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No